accessibility menu, dialog, popup

UserName

CAS RN 185517-21-9

H3C(R)CH3OHO

CAS RN 185517-21-9

IUPAC Name: (2R)-2-propyloctanoic acid
Synonyms: arundic acid acidum arundicum acido arzndico acide arundique
Molecular Weight (g/mol): 186.3
Molecular Formula: C11H22O2
InChi Key: YCYMCMYLORLIJX-SNVBAGLBSA-N
SMILES: CCCCCC[C@@H](CCC)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results