missing translation for 'onlineSavingsMsg'
Learn More

R-(-)-Arundic Acid, TRC

Product Code. 30334187
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30334187

Brand: Toronto Research Chemicals TRCA77950050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-(-)-Arundic acid
CAS 185517-21-9
Molecular Formula C11 H22 O2
Quantity 50 mg
Synonym R-(-)-Arundic acid - Bio-X TM
InChI Formula InChI=1S/C11H22O2/c1-3-5-6-7-9-10(8-4-2)11(12)13/h10H,3-9H2,1-2H3,(H,12,13)/t10-/m1/s1
SMILES CCCCCC[C@@H](CCC)C(=O)O
IUPAC Name (2R)-2-propyloctanoic acid
Molecular Weight (g/mol) 186.29
Formula Weight 186.162
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.