accessibility menu, dialog, popup

UserName

CAS RN 184970-26-1

FBrFFF

CAS RN 184970-26-1

IUPAC Name: 4-(bromomethyl)-1-fluoro-2-(trifluoromethyl)benzene
Synonyms: 4-(bromomethyl)-1-fluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 257.03
Molecular Formula: C8H5BrF4
InChi Key: JVQDTRQMKZMBAU-UHFFFAOYSA-N
SMILES: FC1=CC=C(CBr)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results