accessibility menu, dialog, popup

UserName

FClOF

CAS RN 18355-73-2

IUPAC Name: 2,3-difluorobenzoyl chloride
Synonyms: 2,3-difluorobenzoyl chloride
Molecular Weight (g/mol): 176.55
Molecular Formula: C7H3ClF2O
InChi Key: CDCFERWURRNLLA-UHFFFAOYSA-N
SMILES: FC1=CC=CC(C(Cl)=O)=C1F
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)