missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzoyl chloride, 97%, Thermo Scientific™

Product Code. 11444110
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444110 5 g 5g
11409866 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444110 Supplier Thermo Scientific Alfa Aesar Supplier No. B20333.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18355-73-2
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD00042499
InChI Key CDCFERWURRNLLA-UHFFFAOYSA-N
Synonym 2,3-difluorobenzoylchloride, benzoylchloride, 2,3-difluoro, benzoyl chloride, 2,3-difluoro, pubchem16225, acmc-1cabv, 2,3-difluoro-benzoyl chloride, 2,3-bis fluoranyl benzoyl chloride, 2,3-difluorobenzoyl chloride, 3-chlorocarbonyl-1,2-difluorobenzene
PubChem CID 519549
IUPAC Name 2,3-difluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1)F)F)C(=O)Cl

Specifications

Density 1.423
Boiling Point 189°C to 190°C
Flash Point 90°C (194°F)
Refractive Index 1.5143
Quantity 5 g
UN Number UN3265
Beilstein 2965225
Sensitivity Moisture sensitive
Formula Weight 176.55
Percent Purity 97%
Chemical Name or Material 2,3-Difluorobenzoyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.