CAS RN 1799506-30-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress Cl-C6-PEG4-O-CH2COOH
MedChemExpress Cl-C6-PEG4-O-CH2COOH (PROTAC Linker 4) is a PEG-based PROTAC linker can be used in the synthesis of chloroalkane-containing PROTACs (HaloPROTACs).