CAS RN 179923-32-1
IUPAC Name:
(2,3,4,5-tetrafluorophenyl)boronic acid
Synonyms:
2,3,4,5-tetrafluorophenylboronic acid
Molecular Weight (g/mol):
193.89
Molecular Formula:
C6H3BF4O2
InChi Key:
GXGSBMNUHWAXHP-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(F)=C(F)C(F)=C1F
1
–
1
of
1
results
1
2,3,4,5-Tetrafluorobenzeneboronic acid, 97%, Thermo Scientific™
CAS: 179923-32-1 Molecular Formula: C6H3BF4O2 Molecular Weight (g/mol): 193.892 MDL Number: MFCD04039308 InChI Key: GXGSBMNUHWAXHP-UHFFFAOYSA-N Synonym: 2,3,4,5-tetrafluorophenyl boronic acid,2,3,4,5-tetrafluorobenzeneboronic acid,tetrafluorophenylboronic acid,acmc-1c04s,2,3,4,5-tetrafluorophenylboranic acid,boronic acid, 2,3,4,5-tetrafluorophenyl,boronic acid,b-2,3,4,5-tetrafluorophenyl,2,3,4,5-tetrafluorophenylboronic acid,2,3,4,5-tetrakis fluoranyl phenyl boronic acid,boronic acid, b-2,3,4,5-tetrafluorophenyl PubChem CID: 2783273 IUPAC Name: (2,3,4,5-tetrafluorophenyl)boronic acid SMILES: B(C1=CC(=C(C(=C1F)F)F)F)(O)O