missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrafluorobenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 11389795
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389795 250 mg 250mg
11379795 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389795 Supplier Thermo Scientific Alfa Aesar Supplier No. L19824.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179923-32-1
Molecular Formula C6H3BF4O2
Molecular Weight (g/mol) 193.892
MDL Number MFCD04039308
InChI Key GXGSBMNUHWAXHP-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorophenyl boronic acid, 2,3,4,5-tetrafluorobenzeneboronic acid, tetrafluorophenylboronic acid, acmc-1c04s, 2,3,4,5-tetrafluorophenylboranic acid, boronic acid, 2,3,4,5-tetrafluorophenyl, boronic acid,b-2,3,4,5-tetrafluorophenyl, 2,3,4,5-tetrafluorophenylboronic acid, 2,3,4,5-tetrakis fluoranyl phenyl boronic acid, boronic acid, b-2,3,4,5-tetrafluorophenyl
PubChem CID 2783273
IUPAC Name (2,3,4,5-tetrafluorophenyl)boronic acid
SMILES B(C1=CC(=C(C(=C1F)F)F)F)(O)O

Specifications

Melting Point ∼250°C to 255°C
Quantity 250 mg
Beilstein 8554485
Formula Weight 193.9
Percent Purity 97%
Chemical Name or Material 2,3,4,5-Tetrafluorobenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.