missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Thienyl)ethanohydrazide, 97%, Thermo Scientific™

Product Code. 10692463
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10692463 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10692463 Supplier Thermo Scientific Maybridge Supplier No. SPB02354DA
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 175276-94-5
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H8N2OS
MDL Number MFCD00052946
Quantity 1 g
Synonym 2-thiophen-3-yl acetohydrazide, thiophene-3-acetic acid hydrazide, 2-3-thienyl acetohydrazide, 2-thien-3-yl acetohydrazide, 3-thiopheneacetic acid,hydrazide, 3 thienyl-acetic acid hydrazide, 2-thien-3-ylacetohydrazide, acmc-20ajii, maybridge1_008350, 3-thienyl acetohydrazide
InChI Key YNMRDHMMFCGQBD-UHFFFAOYSA-N
SMILES NNC(=O)CC1=CSC=C1
IUPAC Name 2-(thiophen-3-yl)acetohydrazide
Molecular Weight (g/mol) 156.20
PubChem CID 468649
Formula Weight 156.21
Percent Purity 97%
Chemical Name or Material 2-(3-Thienyl)ethanohydrazide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.