CAS RN 173546-57-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(Z)-Pentatriacont-9-en-18-one, TRC
CAS: 173546-57-1 Molecular Formula: C35H68O Molecular Weight (g/mol): 504.91 IUPAC Name: (Z)-pentatriacont-9-en-18-one SMILES: CCCCCCCCCCCCCCCCCC(=O)CCCCCCC\C=C/CCCCCCCC