missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Pentatriacont-9-en-18-one, TRC

Product Code. 30428767
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428767 5 mg 5mg
30413001 25 mg -
30424915 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428767 Supplier Toronto Research Chemicals Supplier No. TRCP2849955MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 173546-57-1
Molecular Formula C35H68O
Molecular Weight (g/mol) 504.91
IUPAC Name (Z)-pentatriacont-9-en-18-one
SMILES CCCCCCCCCCCCCCCCCC(=O)CCCCCCC\C=C/CCCCCCCC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C35H68O/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35(36)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17,19H,3-16,18,20-34H2,1-2H3/b19-17-
Formula Weight 504.527
Chemical Name or Material (Z)-Pentatriacont-9-en-18-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.