accessibility menu, dialog, popup

UserName

CAS RN 1706428-65-0

CH3OFFFOOHH2N

CAS RN 1706428-65-0

IUPAC Name: 2-amino-3-[2-methyl-4-(trifluoromethoxy)phenyl]propanoic acid
Synonyms: 2-amino-3-[2-methyl-4-(trifluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 263.22
Molecular Formula: C11H12F3NO3
InChi Key: POJFIIFTOPDUEM-UHFFFAOYNA-N
SMILES: CC1=C(CC(N)C(O)=O)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)