CAS RN 164298-27-5
CAS RN 164298-27-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Fluoro-1,3-dimethylimidazolidinium hexafluorophosphate, 97%, Thermo Scientific™
CAS: 164298-27-5 Molecular Formula: C5H10FN2·F6P Molecular Weight (g/mol): 262.11 Synonym: 1, 3-Dimethyl-2-fluoroimidazolidinium hexafluorophosphate,DFIH