missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-1,3-dimethylimidazolidinium hexafluorophosphate, 97%, Thermo Scientific™

Product Code. 11915711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11915711 25 g 25g
11905711 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11915711 Supplier Thermo Scientific Acros Supplier No. 439470250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 164298-27-5
Molecular Formula C5H10FN2·F6P
Molecular Weight (g/mol) 262.11
Synonym 1, 3-Dimethyl-2-fluoroimidazolidinium hexafluorophosphate, DFIH

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 170°C to 172°C
Color White
Elemental Analysis 22.00 to 23.83% C, 10.26 to 11.12% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 25 g
Formula Weight 262.11
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Fluoro-1, 3-dimethylimidazolidinium hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-1,3-dimethylimidazolidinium hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.