missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Isovaleroylglycine, MedChemExpress
Description
N-Isovaleroylglycine is an acyl glycine and could be used as a biomarker for the predispositon for weight gain and obesity.
Specifications
Specifications
| Chemical Name or Material | N-Isovaleroylglycine |
| CAS | 16284-60-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C7H13NO3 |
| Solubility Information | DMSO : 100 mg/mL (628.22 mM; Need ultrasonic) |
| SMILES | O=C(O)CNC(CC(C)C)=O |
| Molecular Weight (g/mol) | 159.18 |
| Quantity | 100 mg |
| Formula Weight | 159.18 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?