CAS RN 1609405-34-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Desmethyl, N-Ethyl Tadalafil, TRC
CAS: 1609405-34-6 Molecular Formula: C23 H21 N3 O4 Molecular Weight (g/mol): 403.4305 Synonym: Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(1,3-benzodioxol-5-yl)-2-ethyl-2,3,6,7,12,12a-hexahydro-, (6R,12aR)-,(6R,12aR)-6-(1,3-Benzodioxol-5-yl)-2-ethyl-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione IUPAC Name: (2R,8R)-2-(1,3-benzodioxol-5-yl)-6-ethyl-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione SMILES: CCN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O