missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl, N-Ethyl Tadalafil, TRC

Product Code. 30382188
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382188 10 mg
30369851 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382188 Supplier Toronto Research Chemicals Supplier No. TRCE76178010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1609405-34-6
Molecular Formula C23 H21 N3 O4
Molecular Weight (g/mol) 403.4305
Synonym Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(1,3-benzodioxol-5-yl)-2-ethyl-2,3,6,7,12,12a-hexahydro-, (6R,12aR)-, (6R,12aR)-6-(1,3-Benzodioxol-5-yl)-2-ethyl-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione
IUPAC Name (2R,8R)-2-(1,3-benzodioxol-5-yl)-6-ethyl-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione
SMILES CCN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H21N3O4/c1-2-25-11-20(27)26-17(23(25)28)10-15-14-5-3-4-6-16(14)24-21(15)22(26)13-7-8-18-19(9-13)30-12-29-18/h3-9,17,22,24H,2,10-12H2,1H3/t17-,22-/m1/s1
Formula Weight 403.1532
Chemical Name or Material Tadalafil-N-ethyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.