CAS RN 16037-91-5
CAS RN 16037-91-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sodium stibogluconate, MedChemExpress
MedChemExpress Sodium stibogluconate (Stibogluconate trisodium nonahydrate) is a potent inhibitor of protein tyrosine phosphatase. Sodium stibogluconate inhibits 99% of SHP-1, SHP-2 and PTP1B activity at 10, 100, 100 μg/mL, respectively.