missing translation for 'onlineSavingsMsg'
Learn More

Sodium stibogluconate, MedChemExpress

Product Code. 16421526
Encompass_Preferred
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
This item is not returnable. View return policy

Product Code. 16421526

Brand: Medchem Express HY100595

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sodium stibogluconate (Stibogluconate trisodium nonahydrate) is a potent inhibitor of protein tyrosine phosphatase. Sodium stibogluconate inhibits 99% of SHP-1, SHP-2 and PTP1B activity at 10, 100, 100 μg/mL, respectively.

Sodium stibogluconate (Stibogluconate trisodium nonahydrate) is a potent inhibitor of protein tyrosine phosphatase. Sodium stibogluconate inhibits 99% of SHP-1, SHP-2 and PTP1B activity at 10, 100, 100 μg/mL, respectively.

Specifications

Chemical Name or Material Sodium stibogluconate
Physical Form Solid
CAS 16037-91-5
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C12H38Na3O26Sb2
For Use With (Application) Cancer-Kinase/protease
Synonym Stibogluconate trisodium nonahydrate
Solubility Information H2O : 3 mg/mL (3.29 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 1 mg/mL (1.10 mM; Need ultrasonic and warming)
SMILES O=[Sb](O[C@]1([H])[C@H](O)CO)(O[C@@H](C(O)=O)[C@H]1O)O[Sb](O[C@]2([H])[C@H](O)CO)(O[C@@H](C(O)=O)[C@H]2O)=O.[9H2O].[3Na]
Molecular Weight (g/mol) 910.9
Formula Weight 910.9
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.