CAS RN 160296-40-2
CAS RN 160296-40-2
IUPAC Name:
tert-butyl 4-(4-fluorobenzoyl)piperidine-1-carboxylate
Synonyms:
tert-butyl 4-(4-fluorobenzoyl)piperidine-1-carboxylate
Molecular Weight (g/mol):
307.37
Molecular Formula:
C17H22FNO3
InChi Key:
ARCRECZRQVSYIP-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)N1CCC(CC1)C(=O)C1=CC=C(F)C=C1
1
–
1
of
1
results