missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(4-fluorobenzoyl)piperidine-1-carboxylate, ≥95%, Thermo Scientific™

Product Code. 10164004
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10164004 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10164004 Supplier Thermo Scientific Maybridge Supplier No. MO07381DA
 Restricted Item
Estimated Shipment: 23-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 160296-40-2
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C17H22FNO3
MDL Number MFCD04114990
Quantity 1 g
Synonym tert-butyl 4-4-fluorobenzoyl piperidine-1-carboxylate, 1-boc-4-4-fluorobenzoyl piperidine, 1-boc-4-4-fluoro-benzoyl-piperidine, 4-4-fluoro-benzoyl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl4-4-fluorobenzoyl piperidine-1-carboxylate, tert-butyl 4-4-fluorophenyl carbonyl piperidine-1-carboxylate, bio-farma bf002994, 4-4-fluorobenzoyl-n-t-butoxycarbonylpiperidine, 4-4-fluorobenzoyl-n-t-butoxylcarbonylpiperidine, 1-t-butoxycarbonyl-4-4-fluorobenzoyl piperidine
InChI Key ARCRECZRQVSYIP-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C(=O)C2=CC=C(C=C2)F
IUPAC Name tert-butyl 4-(4-fluorobenzoyl)piperidine-1-carboxylate
Molecular Weight (g/mol) 307.365
PubChem CID 7016484
Formula Weight 307.36
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-(4-fluorobenzoyl)piperidine-1-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.