CAS RN 158474-72-7
CAS RN 158474-72-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Propyl dihydrojasmonate (Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Propyl dihydrojasmonate (Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Propyl dihydrojasmonate (Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Prohydrojasmon racemate, MedChemExpress
MedChemExpress Prohydrojasmon racemate (n-Propyl dihydrojasmonate) is the racemate of Prohydrojasmon. Prohydrojasmon is a synthesized plant growth regulator.