missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propyl dihydrojasmonate (Mixture of Diastereomers), TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Prohydrojasmon |
| CAS | 158474-72-7 |
| Purity Grade Notes | GC |
| Molecular Formula | C15 H26 O3 |
| Quantity | 250 mg |
| Synonym | Cyclopentaneacetic acid','3-oxo-2-pentyl-','propyl ester |
| InChI Formula | InChI=1S/C15H26O3/c1-3-5-6-7-13-12(8-9-14(13)16)11-15(17)18-10-4-2/h12-13H,3-11H2,1-2H3/t12-,13-/m1/s1 |
| SMILES | CCCCC[C@@H]1[C@@H](CC(=O)OCCC)CCC1=O |
| IUPAC Name | propyl 2-[(1R,2R)-3-oxo-2-pentylcyclopentyl]acetate |
| Molecular Weight (g/mol) | 254.37 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction