accessibility menu, dialog, popup

UserName

CAS RN 157654-67-6

CH3NHCH3OHCl(E/Z)(E/Z)

CAS RN 157654-67-6

IUPAC Name: 3,5-bis[(4-methylphenyl)methylidene]piperidin-4-one hydrochloride
Synonyms: 3,5-bis[(4-methylphenyl)methylidene]piperidin-4-one hydrochloride
Molecular Weight (g/mol): 339.86
Molecular Formula: C21H22ClNO
InChi Key: ZOKZLTXPTLIWOJ-UHFFFAOYSA-N
SMILES: Cl.CC1=CC=C(C=C2CNCC(=CC3=CC=C(C)C=C3)C2=O)C=C1
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)