missing translation for 'onlineSavingsMsg'
Learn More

(S)-SNAP5114, MedChemExpress

Product Code. 16416587
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16416587 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16416587 Supplier Medchem Express Supplier No. HY103504

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-SNAP5114 is a selective GABA transport inhibitor, with IC50 values of 5 μM and 21 μM for hGAT-3 and rGAT-2, respectively. (S)-SNAP5114 is an anticonvulsant drug.

(S)-SNAP5114 is a selective GABA transport inhibitor, with IC50 values of 5 μM and 21 μM for hGAT-3 and rGAT-2, respectively. (S)-SNAP5114 is an anticonvulsant drug.

Specifications

Chemical Name or Material (S)-SNAP5114
CAS 157604-55-2
Color White
Purity Grade Notes Research
Molecular Formula C30H35NO6
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (197.78 mM; Need ultrasonic)
SMILES O=C([C@@H]1CN(CCOC(C2=CC=C(OC)C=C2)(C3=CC=C(OC)C=C3)C4=CC=C(OC)C=C4)CCC1)O
Molecular Weight (g/mol) 505.6
Formula Weight 505.6
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.