accessibility menu, dialog, popup

UserName

CAS RN 155270-99-8

CH3NONH3CNOH3COCH3NH3CO(E)

CAS RN 155270-99-8

IUPAC Name: 8-[(1E)-2-(3,4-dimethoxyphenyl)ethenyl]-1,3-diethyl-7-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: istradefylline
Molecular Weight (g/mol): 384.44
Molecular Formula: C20H24N4O4
InChi Key: IQVRBWUUXZMOPW-PKNBQFBNSA-N
SMILES: CCN1C2=C(N(C)C(\C=C\C3=CC=C(OC)C(OC)=C3)=N2)C(=O)N(CC)C1=O
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (7)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)