CAS RN 152-62-5
CAS RN 152-62-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Dydrogesterone, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
Dydrogesterone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Dydrogesterone, MedChemExpress
MedChemExpress Dydrogesterone is a potent, orally active progestogen indicated in a wide variety of gynaecological conditions related to progesterone deficiency.