missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dydrogesterone, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Dydrogesterone |
| CAS | 152-62-5 |
| Analyte or Component Names | Dydrogesterone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H28 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | (9beta,10alpha)-Pregna-4,6-diene-3,20-dione, 6-Dehydro-9beta,10alpha-progesterone, 6-Dehydroretroprogesterone, 6alpha-Dehydro-retro-4-pregnene-3,20-dione, Diphaston, Dufaston, Gynorest, Hydrogesterone, Prodel, Retrone; |
| InChI Formula | InChI=1S/C21H28O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4-5,12,16-19H,6-11H2,1-3H3/t16-,17+,18-,19+,20+,21+/m0/s1 |
| Solution Type | Neat |
| SMILES | CC(=O)[C@H]1CC[C@H]2[C@@H]3C=CC4=CC(=O)CC[C@@]4(C)[C@@H]3CC[C@]12C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction