CAS RN 150417-90-6
CAS RN 150417-90-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
GC7 Sulfate, MedChemExpress
MedChemExpress GC7 Sulfate is a deoxyhypusine synthase (DHPS) inhibitor.