missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GC7 Sulfate, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
GC7 Sulfate is a deoxyhypusine synthase (DHPS) inhibitor.
Specifications
Specifications
| Chemical Name or Material | GC7 Sulfate |
| Physical Form | Solid |
| CAS | 150417-90-6 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C8H22N4O4S |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | H2O : 3 mg/mL (11.10 mM; Need ultrasonic and warming) |
| SMILES | NC(NCCCCCCCN)=N.O=S(O)(O)=O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction