accessibility menu, dialog, popup

UserName

CAS RN 149771-09-5

H3COONNH2NFFF

CAS RN 149771-09-5

IUPAC Name: ethyl 2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate
Synonyms: ethyl 2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate
Molecular Weight (g/mol): 235.17
Molecular Formula: C8H8F3N3O2
InChi Key: OUACXMUEZBQRBJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(N)N=C1C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results