CAS RN 149709-63-7
CAS RN 149709-63-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2S,4S)-Sacubitril, MedChemExpress
MedChemExpress (2S,4S)-Sacubitril is the impurity of Sacubitril. Sacubitril is a potent NEP inhibitor that can be used for the research of heart failure.