missing translation for 'onlineSavingsMsg'
Learn More

(2S,4S)-Sacubitril, MedChemExpress

Product Code. 16490678
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16490678

Brand: Medchem Express HY78841

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (2S,4S)-Sacubitril is the impurity of Sacubitril. Sacubitril is a potent NEP inhibitor that can be used for the research of heart failure.

(2S,4S)-Sacubitril is the impurity of Sacubitril. Sacubitril is a potent NEP inhibitor that can be used for the research of heart failure.

Specifications

Chemical Name or Material (2S,4S)-Sacubitril
Physical Form Oil
CAS 149709-63-7
Color Colorless
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C24H29NO5
For Use With (Application) COVID-19-immunoregulation
SMILES [H]N(C(CCC(O)=O)=O)[C@H](CC(C=C1)=CC=C1C2=CC=CC=C2)C[C@@H](C(OCC)=O)C
Molecular Weight (g/mol) 411.49
Formula Weight 411.49
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.