CAS RN 1493764-08-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ UNC 2881
Potent Mer kinase inhibitor
2
UNC2881, MedChemExpress
MedChemExpress UNC2881 is a potent and specific Mer kinase inhibitor; inhibits steady-state Mer kinase phosphorylation with an IC50 value of 22 nM.