missing translation for 'onlineSavingsMsg'
Learn More

UNC2881, MedChemExpress

Product Code. 16496838
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496838 10 mg 10mg
16482907 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16496838 Supplier Medchem Express Supplier No. HY15798
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress UNC2881 is a potent and specific Mer kinase inhibitor; inhibits steady-state Mer kinase phosphorylation with an IC50 value of 22 nM.

UNC2881 is a potent and specific Mer kinase inhibitor

Specifications

Chemical Name or Material UNC2881
CAS 1493764-08-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H33N7O2
Quantity 10 mg
Solubility Information DMSO : ≥ 44 mg/mL (94.91 mM)
SMILES CCCCNC1=NC(N[C@H]2CC[C@H](O)CC2)=C(C(NCC3=CC=C(N4C=NC=C4)C=C3)=O)C=N1
Molecular Weight (g/mol) 463.58
Formula Weight 463.58
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.