CAS RN 147591-46-6
CAS RN 147591-46-6
IUPAC Name:
1-(4-methoxyphenyl)-2-methyl-3-nitro-1H-indol-6-ol
Synonyms:
1-(4-methoxyphenyl)-2-methyl-3-nitroindol-6-ol
Molecular Weight (g/mol):
298.3
Molecular Formula:
C16H14N2O4
InChi Key:
VVZNWYXIOADGSW-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)N1C(C)=C(C2=CC=C(O)C=C12)[N+]([O-])=O
1
–
2
of
2
results
1
ID-8, MedChemExpress
MedChemExpress ID-8 is an inhibitor of dual-specificity tyrosine phosphorylation-regulated kinase (DYRK). ID-8 sustains embryonic stem cell (ESC) self-renewal and pluripotency. ID-8 enhances Wnt-mediated hESC survival and proliferation via inhibition of DYRKs.
2