missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies ID-8
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
- ID-8 is an indole derivative (Miyabayashi et al.) that inhibits members of the dual-specificity tyrosine phosphorylation-regulated kinase (DYRK) family (Hasegawa et al.)
- Enables maintenance of mouse embryonic stem (ES) cells in the absence of mouse embryonic fibroblast (MEF) feeder cells, serum, or LIF (Miyabayashi et al.)
- In combination with Wnt, supports human ES cell proliferation and survival, without FGF or TGFβ (Hasegawa et al.)
Specifications
Specifications
| CAS | 147591-46-6 |
| Quantity | 5 mg |
| Molecular Formula | C16H14N2O4 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction