CAS RN 145742-34-3
CAS RN 145742-34-3
IUPAC Name:
4,5-difluoro-2-methoxybenzaldehyde
Synonyms:
4,5-difluoro-2-methoxybenzaldehyde
Molecular Weight (g/mol):
172.13
Molecular Formula:
C8H6F2O2
InChi Key:
ZCLXUHQRYFUSLE-UHFFFAOYSA-N
SMILES:
COC1=CC(F)=C(F)C=C1C=O
1
–
1
of
1
results
1
4,5-Difluoro-2-methoxybenzaldehyde, 97%, Thermo Scientific™
CAS: 145742-34-3 Molecular Formula: C8H6F2O2 Molecular Weight (g/mol): 172.131 MDL Number: MFCD06246884 InChI Key: ZCLXUHQRYFUSLE-UHFFFAOYSA-N Synonym: acmc-1bwm7,intermediates-zcf02113,2-methoxy-4,5-difluorobenzaldehyde,4,5-difluoro-2-methoxy-benzaldehyde,benzaldehyde,4,5-difluoro-2-methoxy,benzaldehyde, 4,5-difluoro-2-methoxy,4,5-difluoro-2-methoxybenzaldehyde,4,5-bis fluoranyl-2-methoxy-benzaldehyde PubChem CID: 15434552 IUPAC Name: 4,5-difluoro-2-methoxybenzaldehyde SMILES: COC1=CC(=C(C=C1C=O)F)F