CAS RN 1456872-74-4
CAS RN 1456872-74-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress PL553
MedChemExpress PL553 is a specific and high-affinity fluorigenic substrate of Leukotriene A4 hydrolase, with a λmax of 210 nm and λem of 410 nm.