CAS RN 145040-37-5
CAS RN 145040-37-5
IUPAC Name:
1-{[(cyclohexyloxy)carbonyl]oxy}ethyl 2-ethoxy-1-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-1,3-benzodiazole-7-carboxylate
Synonyms:
1-{[(cyclohexyloxy)carbonyl]oxy}ethyl 2-ethoxy-3-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-1,3-benzodiazole-4-carboxylate
Molecular Weight (g/mol):
610.67
Molecular Formula:
C33H34N6O6
InChi Key:
GHOSNRCGJFBJIB-UHFFFAOYNA-N
SMILES:
CCOC1=NC2=CC=CC(C(=O)OC(C)OC(=O)OC3CCCCC3)=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1
1
–
4
of
4
results
2
Candesartan Cilexetil, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Candesartan Cilexetil, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
4
Candesartan Cilexetil, MedChemExpress
MedChemExpress Candesartan Cilexetil (TCV-116) is an angiotensin II receptor antagonist used mainly for the treatment of hypertension.