missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Candesartan cilexetil

Product Code. 17251202 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 17251202

Brand: Thermo Scientific Chemicals 466190010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 145040-37-5
Quantity 1 g
Molecular Formula C33H34N6O6
Formula Weight 610.67
InChI Key GHOSNRCGJFBJIB-UHFFFAOYNA-N
SMILES CCOC1=NC2=CC=CC(C(=O)OC(C)OC(=O)OC3CCCCC3)=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1
IUPAC Name 1-{[(cyclohexyloxy)carbonyl]oxy}ethyl 2-ethoxy-1-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-1,3-benzodiazole-7-carboxylate
Molecular Weight (g/mol) 610.67
Chemical Name or Material Candesartan cilexetil
compliance-icons compliance-icons
Product Identifier
  • Candesartan cilexetil
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Candesartan cilexetil

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.