missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Candesartan Cilexetil, MedChemExpress
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
Description
Candesartan Cilexetil (TCV-116) is an angiotensin II receptor antagonist used mainly for the treatment of hypertension.
Specifications
Specifications
| Chemical Name or Material | Candesartan Cilexetil |
| CAS | 145040-37-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C33H34N6O6 |
| Quantity | 500 mg |
| Synonym | TCV-116 |
| Solubility Information | DMSO : ≥ 50 mg/mL (81.88 mM) |
| SMILES | O=C(C1=C2C(N=C(OCC)N2CC3=CC=C(C4=CC=CC=C4C5=NNN=N5)C=C3)=CC=C1)OC(OC(OC6CCCCC6)=O)C |
| Molecular Weight (g/mol) | 610.66 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction