accessibility menu, dialog, popup

UserName

CAS RN 144689-63-4

CH3NNNNHNNOOCH3OOOCH3H3COH

CAS RN 144689-63-4

IUPAC Name: (5-methyl-2-oxo-2H-1,3-dioxol-4-yl)methyl 4-(2-hydroxypropan-2-yl)-2-propyl-1-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-imidazole-5-carboxylate
Synonyms: benicar olmesartan medoxomil olmesartan medoximil olmetec benicar hct votum
Molecular Weight (g/mol): 558.6
Molecular Formula: C29H30N6O6
InChi Key: UQGKUQLKSCSZGY-UHFFFAOYSA-N
SMILES: CCCC1=NC(=C(N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1)C(=O)OCC1=C(C)OC(=O)O1)C(C)(C)O
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)