missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olmesartan Medoxomil, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Olmesartan Medoxomil
Specifications
Specifications
| Chemical Name or Material | Olmesartan medoxomil |
| CAS | 144689-63-4 |
| Analyte or Component Names | Olmesartan medoxomil |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29 H30 N6 O6 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Olmesartan medoxomil, (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 4-(1-hydroxy-1-methylethyl)-2-propyl-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazole-5-carboxylate, 1H-Imidazole-5-carboxylic acid, 4-(1-hydroxy-1-methylethyl)-2-propyl-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl ester, 1H-Imidazole-5-carboxylic acid, 4-(1-hydroxy-1-methylethyl)-2-propyl-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl ester (9CI), 1,3-Dioxole, 1H-imidazole-5-carboxylic acid deriv., Benicar, CS 866, Olmesartan medoxomil, Olmetec |
| InChI Formula | InChI=1S/C29H30N6O6/c1-5-8-23-30-25(29(3,4)38)24(27(36)39-16-22-17(2)40-28(37)41-22)35(23)15-18-11-13-19(14-12-18)20-9-6-7-10-21(20)26-31-33-34-32-26/h6-7,9-14,38H,5,8,15-16H2,1-4H3,(H,31,32,33,34) |
| SMILES | CCCc1nc(c(C(=O)OCC2=C(C)OC(=O)O2)n1Cc3ccc(cc3)c4ccccc4c5nnn[nH]5)C(C)(C)O |
| IUPAC Name | (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl 5-(2-hydroxypropan-2-yl)-2-propyl-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction