accessibility menu, dialog, popup

UserName

CAS RN 142720-24-9

H3CNCH3ONHF

CAS RN 142720-24-9

IUPAC Name: 2-[2-(4-fluorophenyl)-1H-indol-3-yl]-N,N-dihexylacetamide
Synonyms: 2-[2-(4-fluorophenyl)-1H-indol-3-yl]-N,N-dihexylacetamide
Molecular Weight (g/mol): 436.62
Molecular Formula: C28H37FN2O
InChi Key: VUWXAQFLTSBUDB-UHFFFAOYSA-N
SMILES: CCCCCCN(CCCCCC)C(=O)CC1=C(NC2=CC=CC=C12)C1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results