accessibility menu, dialog, popup

UserName

CAS RN 1421263-93-5

H3CNCH3OFF

CAS RN 1421263-93-5

IUPAC Name: 2,6-difluoro-N,N-dipropylbenzamide
Synonyms: 2,6-difluoro-N,N-dipropylbenzamide
Molecular Weight (g/mol): 241.28
Molecular Formula: C13H17F2NO
InChi Key: IUMPPMIJZVBJQU-UHFFFAOYSA-N
SMILES: CCCN(CCC)C(=O)C1=C(F)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results