missing translation for 'onlineSavingsMsg'
Learn More

Royal Jelly acid, MedChemExpress

Product Code. 16443916
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443916 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16443916 Supplier Medchem Express Supplier No. HYN1363

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Royal Jelly acid (Queen Bee Acid) is a fatty acid constituent of royal jelly, promotes the growth and protection of neurons, reduces anxiety-like phenotypes.

Royal Jelly acid (Queen Bee Acid) is a fatty acid constituent of royal jelly, promotes the growth and protection of neurons, reduces anxiety-like phenotypes.

Specifications

Chemical Name or Material Royal Jelly acid
CAS 14113-05-4
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C10H18O3
Quantity 25 mg
Synonym Queen Bee Acid (E)-10-Hydroxy-2-decenoic acid
Solubility Information DMSO : 100 mg/mL (536.91 mM; Need ultrasonic)
SMILES O=C(O)/C=C/CCCCCCCO
Molecular Weight (g/mol) 186.25
Formula Weight 186.25
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.