CAS RN 139968-49-3
CAS RN 139968-49-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Metaflumizone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Metaflumizone, MedChemExpress
MedChemExpress Metaflumizone is a semicarbazone insecticide, acts as a potent sodium channel blocker.