missing translation for 'onlineSavingsMsg'
Learn More

Metaflumizone, MedChemExpress

Product Code. 16481616
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16481616

Brand: Medchem Express HY116448

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Metaflumizone is a semicarbazone insecticide, acts as a potent sodium channel blocker.

Metaflumizone is a semicarbazone insecticide, acts as a potent sodium channel blocker.

Specifications

Chemical Name or Material Metaflumizone
CAS 139968-49-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H16F6N4O2
Quantity 500 mg
Synonym BAS-320I
Solubility Information DMSO : 300 mg/mL (592.42 mM; Need ultrasonic)
SMILES FC(F)(F)C1=CC(/C(CC2=CC=C(C=C2)C#N)=N\NC(NC3=CC=C(C=C3)OC(F)(F)F)=O)=CC=C1
Molecular Weight (g/mol) 506.4
Formula Weight 506.4
Percent Purity 95.12%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.