accessibility menu, dialog, popup

UserName

CAS RN 13735-04-1

SOOHBr

CAS RN 13735-04-1

IUPAC Name: 3-[(4-bromophenyl)sulfanyl]propanoic acid
Synonyms: 3-[(4-bromophenyl)sulfanyl]propanoic acid
Molecular Weight (g/mol): 261.13
Molecular Formula: C9H9BrO2S
InChi Key: CGKAMPCZJIBUGT-UHFFFAOYSA-N
SMILES: OC(=O)CCSC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results